missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate 99.0+%, TCI America™

Catalog No. p-7135764
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001525G 25 g
S0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001525G Supplier TCI America Supplier No. S001525G
Only null left

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -9°C
Color Yellow
Boiling Point 224°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO4725000
TSCA Yes