missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate, FCC, 98%, Spectrum™ Chemical

Catalog No. p-7157752
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604450 500 mL Amber Glass Bottle
18604451 4 L Amber Glass Bottle
18604452 20 L Amber Glass Bottle
18604453 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604450 Supplier Spectrum Chemical Mfg Cor Supplier No. M1363500ML
Only null left

M1363, 119-36-8, C8H8O3

Spectrum™ Chemical Methyl Salicylate, FCC is used as a fragrant in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.535-1.538
Quantity 500 mL
Percent Purity 98%
Grade FCC
Physical Form Liquid