missing translation for 'onlineSavingsMsg'
Learn More

Methyl salicylate, 99%

Catalog No. AC220490050
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220490050 5 mL Glass bottle
AC220495000 500 mL Glass bottle
AC220490010 1 L Glass bottle
AC220490025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC220490050 Supplier Thermo Scientific Chemicals Supplier No. 220490050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119-36-8 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -8°C to 7°C
Color Colorless to Yellow
Density 1.1825g/mL
Boiling Point 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.5380
Linear Formula 2-(HO)C6H4CO2CH3
Quantity 5 mL
Beilstein 10,7
Specific Gravity 1.1825
Merck Index 15,6193
Solubility Information Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C)
Formula Weight 152.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl salicylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 204-317-7

RUO – Research Use Only