missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(-)-lactate, 97%

Catalog No. p-3779154
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213062500 250 g Glass bottle
AC213060010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC213062500 Supplier Thermo Scientific Chemicals Supplier No. 213062500
Only null left

CAS: 27871-49-4 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27871-49-4
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00064265
InChI Key LPEKGGXMPWTOCB-VKHMYHEASA-N
Synonym methyl s---lactate, methyl l-lactate, s-methyl lactate, --methyl l-lactate, s-lactic acid methyl ester, methyl 2s-2-hydroxypropanoate, --methyl lactate, methyl s-2-hydroxypropionate, propanoic acid, 2-hydroxy-, methyl ester, 2s, --lactic acid methyl ester
PubChem CID 94386
ChEBI CHEBI:83222
IUPAC Name methyl (2S)-2-hydroxypropanoate
SMILES COC(=O)[C@H](C)O

Specifications

Acidity 0.2% max. (as lactic acid)
Density 1.0920g/mL
Boiling Point 144.0°C to 145.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4100 to 1.4150
Linear Formula CH3CH(OH)CO2CH3
Quantity 250 g
Beilstein 03, 264
Specific Gravity 1.092
Specific Rotation Condition − 8.50 (20.00°C neat)
Specific Rotation − 8.50
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: miscible with most organic solvents,soluble in alcohol and ether
Viscosity 2.9 mPa.s (25°C)
Formula Weight 104.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-lactate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-704-9

RUO – Research Use Only