missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(+)-3-hydroxybutyrate, 98%

Catalog No. p-7047941
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1999606 5 g
AAL1999614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1999606 Supplier Thermo Scientific Chemicals Supplier No. L1999606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 53562-86-0 | C5H10O3 | 118.13 g/mol

Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (S)-(+)-3-hydroxybutyrate serves as an optically active starting material used in organic synthesis. It is also used in the preparation of (-)-methyl elenolate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids.

Chemical Identifiers

CAS 53562-86-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00064461
InChI Key LDLDJEAVRNAEBW-UHFFFAOYNA-N
Synonym methyl s-+-3-hydroxybutyrate, s-methyl 3-hydroxybutanoate, methyl s-3-hydroxybutyrate, s-3-hydroxybutanoic acid methyl ester, unii-g10281a5ah, methyl 3s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, methyl ester, 3s, methyl-s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid methyl ester, s-3-hydroxybutyric acid methyl ester
PubChem CID 6950307
IUPAC Name methyl (3S)-3-hydroxybutanoate
SMILES COC(=O)CC(C)O

Specifications

Density 1.055
Boiling Point 56°C to 58°C (11 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.422
Quantity 5 g
Beilstein 1720583
Solubility Information Miscible with alcohol. Immiscible with water.
Optical Rotation +48.5° (c=1.3 in Chloroform)
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl (S)-(+)-3-hydroxybutyrate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H318-H227-H315
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 258-628-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only