accessibility menu, dialog, popup

UserName

H3COOCH3OH

CAS RN 53562-86-0

IUPAC Name: methyl 3-hydroxybutanoate
Synonyms: methyl 3-hydroxybutyrate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: LDLDJEAVRNAEBW-UHFFFAOYNA-N
SMILES: COC(=O)CC(C)O
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)