missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(-)-3-Bromoisobutyrate 97.0+%, TCI America™

Catalog No. B21401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21401G 1 g
B21405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21401G Supplier TCI America Supplier No. B21401G
Only null left

Chemical Identifiers

CAS 98190-85-3
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00064487
InChI Key FKWNAVCXZSQYTA-UHFFFAOYNA-N
Synonym (S)-(-)-3-Bromo-2-methylpropionic Acid Methyl Ester, (S)-(-)-3-Bromoisobutyric Acid Methyl Ester, Methyl (S)-(-)-3-Bromo-2-methylpropionate
PubChem CID 5479124
IUPAC Name methyl 3-bromo-2-methylpropanoate
SMILES COC(=O)C(C)CBr

Specifications

Color Yellow
Quantity 1 g
Formula Weight 181.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-3-Bromoisobutyrate