accessibility menu, dialog, popup

UserName

CH3BrOOCH3

CAS RN 98190-85-3

IUPAC Name: methyl 3-bromo-2-methylpropanoate
Synonyms: methyl 3-bromo-2-methylpropanoate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: FKWNAVCXZSQYTA-UHFFFAOYNA-N
SMILES: COC(=O)C(C)CBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results