missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(-)-2-Isocyanatopropionate 98.0+%, TCI America™

Catalog No. I04655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04655G 5 g
I04651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04655G Supplier TCI America Supplier No. I04655G

Chemical Identifiers

CAS 30293-82-4
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00191530
InChI Key ZYILYULSVKLHKZ-BYPYZUCNSA-N
Synonym (S)-(-)-2-Isocyanatopropionic Acid Methyl Ester
PubChem CID 7019162
IUPAC Name methyl (2S)-2-isocyanatopropanoate
SMILES CC(C(=O)OC)N=C=O

Specifications

Color Yellow
Boiling Point 61°C
Quantity 5 g
UN Number 2206
Formula Weight 129.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Isocyanatopropionate