missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate 98.0+%, TCI America™

Catalog No. I04665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04665G 5 g
I04661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04665G Supplier TCI America Supplier No. I04665G

Chemical Identifiers

CAS 30293-86-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD00191531
InChI Key JCVZXXSBCISMLJ-LURJTMIESA-N
Synonym (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate
PubChem CID 10397066
IUPAC Name methyl (2S)-2-isocyanato-3-methylbutanoate
SMILES CC(C)C(C(=O)OC)N=C=O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 5 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate