missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%
Description
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents
Solubility
Soluble in dichloromethane.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents
Solubility
Soluble in dichloromethane.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 68°C to 72°C |
| Quantity | 1 g |
| UN Number | UN3077 |
| Solubility Information | Soluble in dichloromethane. |
| Optical Rotation | −32° (c=0.5 in Chloroform) |
| Formula Weight | 243.26 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H319-H317 Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only