missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%

Catalog No. AAH6209103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6209103 1 g
AAH6209106 5 g
AAH6209114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6209103 Supplier Thermo Scientific Chemicals Supplier No. H6209103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108963-96-8 | C11H17NO5 | 243.259 g/mol

Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate in medicine, pharmacy, cosmetics, and industrial, artificial flavoring and fragrance agents

Solubility
Soluble in dichloromethane.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 108963-96-8
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD06809720
InChI Key FNTAOUUEQHKLIU-ZETCQYMHSA-N
Synonym boc-l-pyroglutamic acid methyl ester, boc-pyr-ome, s-1-tert-butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate, methyl l-n-boc-pyroglutamate, 1-tert-butyl 2-methyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, methyl boc-l-pyroglutamate, s-5-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, 1-tert-butyl 2-methyl s-5-oxopyrrolidine-1,2-dicarboxylate, boc-pglu-ome, pubchem12135
PubChem CID 10868485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OC

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
UN Number UN3077
Solubility Information Soluble in dichloromethane.
Optical Rotation −32° (c=0.5 in Chloroform)
Formula Weight 243.26
Percent Purity 98%
Chemical Name or Material Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only