missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, Sodium Salt, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5391556
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2003489 10 g Wide Mouth Amber Glass Bottle
2003490 25 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2003489 Supplier Avantor Supplier No. R08602
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym C.I. 13020
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Formula Weight 291.29
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 10 g
Chemical Name or Material Methyl Red, Sodium Salt