missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Sodium Salt, ACS, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Color | Red to Violet |
| pH | 4.2 to 6.2 |
| Quantity | 5 g |
| Grade | ACS |
| Chemical Name or Material | Methyl Red Sodium Salt |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.