missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red sodium salt

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol

Supplier:  Thermo Scientific Chemicals A1745522

 View more versions of this product

Catalog No. AAA1745522


Only null left

Description

Description

Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

Solubility
Soluble in water (800 mg/ml at 25°C), and slightly soluble in ethanol at (25°C).

Notes
Store at room temperature. Reactive with oxidizing agents.
Specifications

Specifications

Methyl Red sodium salt
C15H14N3NaO2
4065488
Soluble in water (800mg/ml at 25°C),and slightly soluble in ethanol at (25°C).
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
291.286
291.29
845-10-3
MFCD00002426
C.I. 13020; Methyl Red, water soluble
GNTPCYMJCJNRQB-UHFFFAOYSA-M
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
4465632
100 g

Chemical Identifiers

845-10-3
291.286
GNTPCYMJCJNRQB-UHFFFAOYSA-M
4465632
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
C15H14N3NaO2
MFCD00002426
C.I. 13020; Methyl Red, water soluble
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-682-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only