missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red, Neutral, Reagent, ACS, Spectrum™ Chemical
M1285, 493-52-7, C15H15N3O2
$350.18 - $17,834.18
Chemical Identifiers
| CAS | 493-52-7 |
|---|---|
| Molecular Formula | C15H15N3O2 |
| Molecular Weight (g/mol) | 269.30 |
| InChI Key | CEQFOVLGLXCDCX-UHFFFAOYSA-N |
| IUPAC Name | 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18604444
|
N/A | 25 g | Amber Glass Bottle |
Each for $350.18
|
|
||||
|
18604445
|
N/A | 100 g | Amber Glass Bottle |
Each for $964.71
|
|
||||
|
18604446
|
N/A | 1 kg | Amber Glass Bottle |
Each for $4,989.79
|
|
||||
|
18604447
|
N/A | 5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Methyl Red, Neutral, Reagent, ACS , also known as C.I. Acid Red 2, is an azo dye that turns red in acidic solutions. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 493-52-7 | |
| 269.30 | |
| 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid |
| C15H15N3O2 | |
| CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O |
Specifications
| 493-52-7 | |
| C15H15N3O2 | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O | |
| 269.30 | |
| Red to Violet |
| 100% | |
| CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
| 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid | |
| ACS | |
| Methyl Red |