missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, Neutral, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18604444
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604444 25 g Amber Glass Bottle
18604445 100 g Amber Glass Bottle
18604446 1 kg Amber Glass Bottle
18604447 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604444 Supplier Spectrum Chemical Mfg Cor Supplier No. M128525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

M1285, 493-52-7, C15H15N3O2

Spectrum™ Chemical Methyl Red, Neutral, Reagent, ACS , also known as C.I. Acid Red 2, is an azo dye that turns red in acidic solutions. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O

Specifications

CAS Max % 100%
Grade ACS
Packaging Amber Glass Bottle
Color Red to Violet
Quantity 25 g
Chemical Name or Material Methyl Red