missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Indicator, Reag. Ph. Eur., Honeywell Fluka™
Reag. Ph. Eur., acid-base indicator
$113.54 - $277.20
Chemical Identifiers
| CAS | 493-52-7 |
|---|---|
| Molecular Formula | C15H15N3O2 |
| Molecular Weight (g/mol) | 269.304 |
| MDL Number | MFCD00002425 |
| InChI Key | CEQFOVLGLXCDCX-UHFFFAOYSA-N |
| Synonym | Acid Red 2 |
| PubChem CID | 10303 |
| IUPAC Name | 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| C15H15N3O2 | |
| MFCD00002425 | |
| Acid Red 2 | |
| 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid |
Specifications
| 179°C to 182°C | |
| C15H15N3O2 | |
| MFCD00002425 | |
| 750102 | |
| Slightly soluble in water | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O | |
| 269.304 | |
| 269.3g/mol | |
| 4.4 to 6.0 | |
| Violet | |
| Methyl Red, Reag. Ph. Eur. |
| 493-52-7 | |
| (CH3)2NC6H4N=NC6H4CO2H | |
| NONH for all modes of transport | |
| Acid Red 2 | |
| CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
| 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid | |
| 10303 | |
| Complies for sensitivity | |
| 0.989g/cm3 | |
| Powder |
Safety and Handling
P273-P391-P501
H411
EINECSNumber : 207-776-1
RTECSNumber : DG8960000