missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, Reag. Ph. Eur., Solstice

Catalog No. 60046144
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046144 25 g Glass Bottle
60046143 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046144 Supplier Solstice Supplier No. 3265425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reag. Ph. Eur., acid-base indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 179°C to 182°C
Packaging Glass Bottle
Suitability Complies for sensitivity
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
Visual Transition Interval 4.4 to 6.0
UN Number NONH for all modes of transport
Beilstein 750102
Density 0.989g/cm3
Color Violet
Solubility Information Slightly soluble in water
Quantity 25 g
Physical Form Powder
Formula Weight 269.3g/mol
Chemical Name or Material Methyl Red, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P273-P391-P501
Precautionary Statement H411
EINECSNumber 207-776-1
RTECSNumber DG8960000