missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, ACS reagent

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

$53.84 - $212.19

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC423970100
View Documents Promotion Details
Thermo Scientific Chemicals
423970100
10 g Glass bottle
Each for $53.84
Case of 6 Each for $0.00
Only null left
 
AC423970250
View Documents Promotion Details
Thermo Scientific Chemicals
423970250
25 g Glass bottle
Each for $75.82
Case of 6 Each for $0.00
Only null left
 
AC423971000
View Documents Promotion Details
Thermo Scientific Chemicals
423971000
100 g Glass bottle
Each for $212.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

493-52-7
269.304
CEQFOVLGLXCDCX-UHFFFAOYSA-N
10303
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
C15H15N3O2
MFCD00002425
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid

Specifications

178.0°C to 182.0°C
Passes Test
(CH3)2NC6H4N=NC6H4CO2H
16, 329
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
CEQFOVLGLXCDCX-UHFFFAOYSA-N
Authentic
269.304
269.3
from pH 4.2 (pink) to pH 6.2 (yellow)
Powder
493-52-7
C15H15N3O2
MFCD00002425
15, 6192
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
10303
ACS Reagent
Purple to Red
Methyl Red, Pure
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only