missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, ACS reagent

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

Supplier:  Thermo Scientific Chemicals 423970250

Quantity 25 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC423970250



Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

493-52-7
269.304
CEQFOVLGLXCDCX-UHFFFAOYSA-N
10303
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
C15H15N3O2
MFCD00002425
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid

Specifications

Methyl Red
Pure
Passes Test
(CH3)2NC6H4N=NC6H4CO2H
16, 329
Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid, C.I. 13020
CEQFOVLGLXCDCX-UHFFFAOYSA-N
Authentic
269.304
269.3
Glass bottle
Purple to Red
Powder
178.0°C to 182.0°C
493-52-7
C15H15N3O2
MFCD00002425
15, 6192
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol, acetic acid
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
10303
ACS Reagent
from pH 4.2 (pink) to pH 6.2 (yellow)
25 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions