missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, 0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™

Supplier:  LabChem LC171007

 View more versions of this product

Catalog No. LC171007


Only null left

Description

Description

Specifications

Specifications

Methyl Red
0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow
99.98,0.02
CH4O
MFCD00004595
Passes Test
CO
32.04
Certified
0.79g/mL
0.79g/mL
125 mL
Liquid
-98°C
65°C
845-10-3
C15H14N3O2Na
Soluble in water
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
291.28
Poly Bottle
Carbon monoxide; Carbon dioxide; May release flammable gases
Orange
9.7°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions