missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylate 98.0+%, TCI America™

Catalog No. p-7127368
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25625G 5 g
D256225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25625G Supplier TCI America Supplier No. D25625G

Chemical Identifiers

CAS 52373-72-5
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00066527
InChI Key DOWWCCDWPKGNGX-RXMQYKEDSA-N
Synonym (+)-2,3-O-Isopropylidene-D-glyceric Acid Methyl Ester, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, (+)-Methyl 2,3-O-Isopropylidene-D-glycerate
PubChem CID 853166
IUPAC Name methyl (4R)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
SMILES CC1(OCC(O1)C(=O)OC)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylate