accessibility menu, dialog, popup

UserName

CAS RN 52373-72-5

IUPAC Name: methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate
Synonyms: methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: DOWWCCDWPKGNGX-UHFFFAOYNA-N
SMILES: COC(=O)C1COC(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results