Learn More
Methyl pyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals
$41.12 - $123.58
Chemical Identifiers
| CAS | 1193-62-0 |
|---|---|
| Molecular Formula | C6H7NO2 |
| Molecular Weight (g/mol) | 125.127 |
| MDL Number | MFCD00817048 |
| InChI Key | VONGYFFEWFJHNP-UHFFFAOYSA-N |
| Synonym | methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1 |
| PubChem CID | 136930 |
| IUPAC Name | methyl 1H-pyrrole-2-carboxylate |
| SMILES | COC(=O)C1=CC=CN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5943703
|
Thermo Scientific Chemicals
H5943703 |
1 g |
Each for $41.12
|
|
|||||
|
AAH5943706
|
Thermo Scientific Chemicals
H5943706 |
5 g |
Each for $123.58
|
|
|||||
Description
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.Â
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.Â
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Chemical Identifiers
| 1193-62-0 | |
| 125.127 | |
| VONGYFFEWFJHNP-UHFFFAOYSA-N | |
| 136930 | |
| COC(=O)C1=CC=CN1 |
| C6H7NO2 | |
| MFCD00817048 | |
| methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1 | |
| methyl 1H-pyrrole-2-carboxylate |
Specifications
| 1193-62-0 | |
| C6H7NO2 | |
| 1 g | |
| Soluble in water. | |
| COC(=O)C1=CC=CN1 | |
| 125.127 | |
| 125.13 | |
| Methyl pyrrole-2-carboxylate |
| 74°C to 78°C | |
| MFCD00817048 | |
| methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1 | |
| VONGYFFEWFJHNP-UHFFFAOYSA-N | |
| methyl 1H-pyrrole-2-carboxylate | |
| 136930 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only