missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Propyl Ether 95.0+%, TCI America™
$180.81 - $2261.10
Chemical Identifiers
| CAS | 557-17-5 |
|---|---|
| Molecular Formula | C4H10O |
| Molecular Weight (g/mol) | 74.12 |
| MDL Number | MFCD00059399 |
| InChI Key | VNKYTQGIUYNRMY-UHFFFAOYSA-N |
| Synonym | 1-Methoxypropane |
| PubChem CID | 11182 |
| IUPAC Name | 1-methoxypropane |
| SMILES | CCCOC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M051025ML
|
TCI America
M051025ML |
25 mL |
Each for $180.81
|
|
|||||
|
M0510500ML
|
TCI America
M0510500ML |
500 mL |
Each for $2,261.10
|
|
|||||
Chemical Identifiers
| 557-17-5 | |
| 74.12 | |
| VNKYTQGIUYNRMY-UHFFFAOYSA-N | |
| 11182 | |
| CCCOC |
| C4H10O | |
| MFCD00059399 | |
| 1-Methoxypropane | |
| 1-methoxypropane |
Specifications
| 557-17-5 | |
| 39°C | |
| MFCD00059399 | |
| 2612 | |
| VNKYTQGIUYNRMY-UHFFFAOYSA-N | |
| 1-methoxypropane | |
| 11182 | |
| ≥95.0% (GC) | |
| Methyl Propyl Ether |
| Colorless | |
| C4H10O | |
| 25 mL | |
| 1-Methoxypropane | |
| CCCOC | |
| 74.12 | |
| 74.12 | |
| Liquid |
Safety and Handling
RTECSNumber : KO2280000
TSCA : No