accessibility menu, dialog, popup

UserName

CAS RN 557-17-5

CH3OH3C

CAS RN 557-17-5

IUPAC Name: 1-methoxypropane
Synonyms: methoxypropane
Molecular Weight (g/mol): 74.12
Molecular Formula: C4H10O
InChi Key: VNKYTQGIUYNRMY-UHFFFAOYSA-N
SMILES: CCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results