missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propyl Ether 95.0+%, TCI America™

Catalog No. M0510500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0510500ML 500 mL
M051025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0510500ML Supplier TCI America Supplier No. M0510500ML
Only null left

Chemical Identifiers

CAS 557-17-5
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00059399
InChI Key VNKYTQGIUYNRMY-UHFFFAOYSA-N
Synonym 1-Methoxypropane
PubChem CID 11182
IUPAC Name 1-methoxypropane
SMILES CCCOC

Specifications

Color Colorless
Boiling Point 39°C
Quantity 500 mL
UN Number 2612
Formula Weight 74.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Propyl Ether
RTECSNumber KO2280000
TSCA No