missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Propyl Disulfide 97.0+%, TCI America™
$55.92 - $130.48
Chemical Identifiers
| CAS | 2179-60-4 |
|---|---|
| Molecular Formula | C4H10S2 |
| Molecular Weight (g/mol) | 122.244 |
| MDL Number | MFCD00013400 |
| InChI Key | PUCHCUYBORIUSM-UHFFFAOYSA-N |
| Synonym | methyl propyl disulfide, disulfide, methyl propyl, 1-methyldisulfanyl propane, 2,3-dithiahexane, methyl propyl disulphide, methyldithiopropane, propyl methyl disulfide, methyl n-propyl disulfide, unii-8m12k3fu0t, methylpropyldisulfide |
| PubChem CID | 16592 |
| IUPAC Name | 1-(methyldisulfanyl)propane |
| SMILES | CCCSSC |
Chemical Identifiers
| 2179-60-4 | |
| 122.244 | |
| PUCHCUYBORIUSM-UHFFFAOYSA-N | |
| 16592 | |
| CCCSSC |
| C4H10S2 | |
| MFCD00013400 | |
| methyl propyl disulfide, disulfide, methyl propyl, 1-methyldisulfanyl propane, 2,3-dithiahexane, methyl propyl disulphide, methyldithiopropane, propyl methyl disulfide, methyl n-propyl disulfide, unii-8m12k3fu0t, methylpropyldisulfide | |
| 1-(methyldisulfanyl)propane |
Specifications
| 2179-60-4 | |
| 71°C | |
| MFCD00013400 | |
| 1993 | |
| PUCHCUYBORIUSM-UHFFFAOYSA-N | |
| 1-(methyldisulfanyl)propane | |
| 16592 | |
| ≥97.0% (GC) | |
| Methyl Propyl Disulfide |
| Yellow | |
| C4H10S2 | |
| 5 g | |
| methyl propyl disulfide, disulfide, methyl propyl, 1-methyldisulfanyl propane, 2,3-dithiahexane, methyl propyl disulphide, methyldithiopropane, propyl methyl disulfide, methyl n-propyl disulfide, unii-8m12k3fu0t, methylpropyldisulfide | |
| CCCSSC | |
| 122.244 | |
| 122.24 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0477
TSCA : Yes