missing translation for 'onlineSavingsMsg'
Learn More

Methyl propionylacetate, 99%

Catalog No. AAB2484814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2484814 25 g
AAB2484822 100 g
AAB2484836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2484814 Supplier Thermo Scientific Chemicals Supplier No. B2484814

CAS: 30414-53-0 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30414-53-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00011705
InChI Key XJMIXEAZMCTAGH-UHFFFAOYSA-N
Synonym methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
PubChem CID 121699
IUPAC Name methyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OC

Specifications

Density 1.037
Boiling Point 73°C to 74°C (5 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.422
Quantity 25 g
Beilstein 1753684
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material Methyl propionylacetate
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 250-184-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only