Learn More
Methyl propionate, 99%
CAS: 554-12-1 | C4H8O2 | 88.106 g/mol
Supplier: Thermo Scientific Chemicals A149440F
Description
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.
Solubility
Soluble in water (5g/L).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Methyl propionate | |
| -88°C | |
| 78°C to 80°C | |
| 99% | |
| 1.377 | |
| 2500 mL | |
| 1737628 | |
| methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural | |
| RJUFJBKOKNCXHH-UHFFFAOYSA-N | |
| methyl propanoate | |
| 11124 | |
| 99% |
| 554-12-1 | |
| 0.915 | |
| −2°C (28°F) | |
| C4H8O2 | |
| MFCD00009306 | |
| UN1248 | |
| 14,6112 | |
| Soluble in water (5g/L). | |
| CCC(=O)OC | |
| 88.106 | |
| 88.11 | |
| Liquid |
Chemical Identifiers
| 554-12-1 | |
| 88.106 | |
| RJUFJBKOKNCXHH-UHFFFAOYSA-N | |
| 11124 | |
| CCC(=O)OC |
| C4H8O2 | |
| MFCD00009306 | |
| methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural | |
| methyl propanoate |
Safety and Handling
GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPIONATE
EINECSNumber : 209-060-4
RTECSNumber : UF5970000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only