missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Propiolate 98.0+%, TCI America™
$74.56 - $301.98
Chemical Identifiers
| CAS | 922-67-8 |
|---|---|
| Molecular Formula | C4H4O2 |
| Molecular Weight (g/mol) | 84.074 |
| MDL Number | MFCD00008572 |
| InChI Key | IMAKHNTVDGLIRY-UHFFFAOYSA-N |
| Synonym | methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester |
| PubChem CID | 13536 |
| IUPAC Name | methyl prop-2-ynoate |
| SMILES | COC(=O)C#C |
Chemical Identifiers
| 922-67-8 | |
| 84.074 | |
| IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
| 13536 | |
| COC(=O)C#C |
| C4H4O2 | |
| MFCD00008572 | |
| methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
| methyl prop-2-ynoate |
Specifications
| 922-67-8 | |
| 102°C | |
| MFCD00008572 | |
| 3272 | |
| IMAKHNTVDGLIRY-UHFFFAOYSA-N | |
| methyl prop-2-ynoate | |
| 13536 | |
| ≥98.0% (GC) | |
| Methyl Propiolate |
| Yellow | |
| C4H4O2 | |
| 5 mL | |
| methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester | |
| COC(=O)C#C | |
| 84.074 | |
| 84.07 | |
| Liquid |
Safety and Handling
RTECSNumber : UE0050000
TSCA : No