accessibility menu, dialog, popup

UserName

CH3OOHC

CAS RN 922-67-8

IUPAC Name: methyl prop-2-ynoate
Synonyms: propiolic acid, methyl ester
Molecular Weight (g/mol): 84.07
Molecular Formula: C4H4O2
InChi Key: IMAKHNTVDGLIRY-UHFFFAOYSA-N
SMILES: COC(=O)C#C
Boiling Point 
  • (2)
  • (6)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)