missing translation for 'onlineSavingsMsg'
Learn More

Methyl propiolate, 99%

CAS: 922-67-8 | C4H4O2 | 84.074 g/mol

Supplier:  Thermo Scientific Chemicals A1474314

 View more versions of this product

Catalog No. AAA1474314


Only null left
Explore available promotions

Description

Description

Methyl propiolate was used in the synthesis of polysubstituted 3-arylaminoacrylate and tetrahydropyrimidin-2-one derivatives. It was also used as a thiol derivatizing agent for capillary electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl propiolate was used in the synthesis of polysubstituted 3-arylaminoacrylate and tetrahydropyrimidin-2-one derivatives. It was also used as a thiol derivatizing agent for capillary electrophoresis.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl propiolate
1.016
10°C (50°F)
C4H4O2
MFCD00008572
UN3272
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
IMAKHNTVDGLIRY-UHFFFAOYSA-N
methyl prop-2-ynoate
13536
99%
922-67-8
98°C to 101°C
Pungent
1.408
25 g
605462
Not miscible in water.
COC(=O)C#C
84.074
84.07

Chemical Identifiers

922-67-8
84.074
IMAKHNTVDGLIRY-UHFFFAOYSA-N
13536
COC(=O)C#C
C4H4O2
MFCD00008572
methyl propiolate, methyl propynoate, 2-propynoic acid, methyl ester, propiolic acid, methyl ester, methyl acetylenecarboxylate, methyl ethynecarboxylate, propynoic acid, methyl ester, propynoic acid methyl ester, methyl propargylate, acetylenecarboxylic acid methyl ester
methyl prop-2-ynoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 213-083-5

RTECSNumber : UE0050000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only