missing translation for 'onlineSavingsMsg'
Learn More

Methyl pivaloylacetate, 98%, Thermo Scientific™

Numéro de catalogue. AC414190010
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC414190010 1g
AC414190100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC414190010 Fournisseur Thermo Scientific Chemicals Code fournisseur 414190010

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 55107-14-7
Formule moléculaire C8H14O3
Poids moléculaire (g/mol) 158.2
Clé InChI XTXCFTMJPRXBBC-UHFFFAOYSA-N
Synonyme methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
PubChem CID 99597
Nom de l’IUPAC methyl 4,4-dimethyl-3-oxopentanoate
SOURIRES CC(C)(C)C(=O)CC(=O)OC

Spécifications

Nom ou substance chimique Methyl pivaloylacetate
Densité 0.9800g/mL
Point d’ébullition 67.0°C to 70.0°C (13.0mmHg)
Point d’éclair 78°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (GC)
Emballage Glass Bottle
Indice de réfraction 1.4300 to 1.4320
Formule linéaire (CH3)3CCOCH2COOCH3
Quantité 1g
Gravité spécifique 0.98
Poids de formule 158.2
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-481-2

RUO – Research Use Only