missing translation for 'onlineSavingsMsg'
Learn More

Methyl (Phenylthio)acetate, TCI America™

Catalog No. P13345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13345G Supplier TCI America Supplier No. P13345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17277-58-6
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00039805
InChI Key MUNSXQQODXYRKI-UHFFFAOYSA-N
Synonym methyl phenylthio acetate, methyl 2-phenylthio acetate, methyl phenylsulfanyl acetate, phenylthio acetic acid methyl ester, methyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, methyl ester, methyl 2-phenylthioacetate, pubchem10851
PubChem CID 123375
IUPAC Name methyl 2-phenylsulfanylacetate
SMILES COC(=O)CSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 182.24
Physical Form Liquid
Chemical Name or Material Methyl (Phenylthio)acetate