missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylpropiolate, 97%

CAS: 4891-38-7 | C10H8O2 | 160.172 g/mol

$120.56 - $1,140.38

Chemical Identifiers

CAS 4891-38-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00041685
InChI Key JFGWPXKGINUNDH-UHFFFAOYSA-N
Synonym methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
PubChem CID 138378
IUPAC Name methyl 3-phenylprop-2-ynoate
SMILES COC(=O)C#CC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2458706
View Documents
Thermo Scientific Chemicals
B2458706
5 g N/A
Only null left
 
AAB2458714
View Documents
Thermo Scientific Chemicals
B2458714
25 g N/A
Only null left
 
AAB2458722
View Documents
Thermo Scientific Chemicals
B2458722
100 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4891-38-7
160.172
JFGWPXKGINUNDH-UHFFFAOYSA-N
138378
COC(=O)C#CC1=CC=CC=C1
C10H8O2
MFCD00041685
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
methyl 3-phenylprop-2-ynoate

Specifications

4891-38-7
109°C to 110°C (2 mmHg)
C10H8O2
1.562
5 g
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
COC(=O)C#CC1=CC=CC=C1
160.172
160.17
Methyl phenylpropiolate
1.083
>110°C (230°F)
C6H5C≡CCO2CH3
MFCD00041685
472813
JFGWPXKGINUNDH-UHFFFAOYSA-N
methyl 3-phenylprop-2-ynoate
138378
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only