missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl phenylpropiolate, 97%
CAS: 4891-38-7 | C10H8O2 | 160.172 g/mol
Supplier: Thermo Scientific Chemicals B2458714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl phenylpropiolate | |
| 1.083 | |
| >110°C (230°F) | |
| C6H5C≡CCO2CH3 | |
| MFCD00041685 | |
| 472813 | |
| JFGWPXKGINUNDH-UHFFFAOYSA-N | |
| methyl 3-phenylprop-2-ynoate | |
| 138378 | |
| 97% |
| 4891-38-7 | |
| 109°C to 110°C (2 mmHg) | |
| C10H8O2 | |
| 1.562 | |
| 25 g | |
| methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
| COC(=O)C#CC1=CC=CC=C1 | |
| 160.172 | |
| 160.17 |
Chemical Identifiers
| 4891-38-7 | |
| 160.172 | |
| JFGWPXKGINUNDH-UHFFFAOYSA-N | |
| 138378 | |
| COC(=O)C#CC1=CC=CC=C1 |
| C10H8O2 | |
| MFCD00041685 | |
| methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate | |
| methyl 3-phenylprop-2-ynoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only