missing translation for 'onlineSavingsMsg'
Learn More

Methyl Phenyl Sulfoxide 98.0+%, TCI America™

Catalog No. M11485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11485G 5 g
M114825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11485G Supplier TCI America Supplier No. M11485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Phenyl Sulfoxide