missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl sulfoxide, 98+%

Catalog No. AAA1500906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500906 5 g
AAA1500914 25 g
AAA1500922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500906 Supplier Thermo Scientific Chemicals Supplier No. A1500906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1193-82-4 | C7H8OS | 140.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1193-82-4
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00002088
InChI Key JXTGICXCHWMCPM-UHFFFAOYSA-N
Synonym methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
PubChem CID 14516
IUPAC Name methylsulfinylbenzene
SMILES CS(=O)C1=CC=CC=C1

Specifications

Melting Point 26°C to 32°C
Boiling Point 93°C to 94°C (0.3 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.5775
Quantity 5 g
Beilstein 1904976
Sensitivity Hygroscopic
Formula Weight 140.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfoxide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-781-2
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only