missing translation for 'onlineSavingsMsg'
Learn More

Methyl (Phenyl 2,3,4-Tri-O-acetyl-1-thio-beta-D-glucopyranosid)uronate 98.0+%, TCI America™

Catalog No. M17591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17591G Supplier TCI America Supplier No. M17591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62812-42-4
Molecular Formula C19H22O9S
Molecular Weight (g/mol) 426.436
InChI Key AXUXNCRKZCBMKR-CWLGOENISA-N
Synonym GlcA[234Ac,6Me]-beta-SPh
PubChem CID 15507916
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxane-2-carboxylate
SMILES CC(=O)OC1C(C(OC(C1OC(=O)C)SC2=CC=CC=C2)C(=O)OC)OC(=O)C

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 426.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (Phenyl 2,3,4-Tri-O-acetyl-1-thio-beta-D-glucopyranosid)uronate
TSCA No
Recommended Storage Refrigerator