CAS RN 62812-42-4
IUPAC Name:
methyl 3,4,5-tris(acetyloxy)-6-(phenylsulfanyl)oxane-2-carboxylate
Synonyms:
methyl 3,4,5-tris(acetyloxy)-6-(phenylsulfanyl)oxane-2-carboxylate
Molecular Weight (g/mol):
426.44
Molecular Formula:
C19H22O9S
InChi Key:
AXUXNCRKZCBMKR-UHFFFAOYNA-N
SMILES:
COC(=O)C1OC(SC2=CC=CC=C2)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
Methyl (Phenyl 2,3,4-Tri-O-acetyl-1-thio-beta-D-glucopyranosid)uronate 98.0+%, TCI America™
CAS: 62812-42-4 Molecular Formula: C19H22O9S Molecular Weight (g/mol): 426.436 InChI Key: AXUXNCRKZCBMKR-CWLGOENISA-N Synonym: GlcA[234Ac,6Me]-beta-SPh PubChem CID: 15507916 IUPAC Name: methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxane-2-carboxylate SMILES: CC(=O)OC1C(C(OC(C1OC(=O)C)SC2=CC=CC=C2)C(=O)OC)OC(=O)C