missing translation for 'onlineSavingsMsg'
Learn More

Methyl Phenoxyacetate 99.0+%, TCI America™

Catalog No. P0110500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0110500G 500 g
P011025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0110500G Supplier TCI America Supplier No. P0110500G
Only null left

Chemical Identifiers

CAS 2065-23-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00010227
InChI Key BZCKRPHEZOHHBK-UHFFFAOYSA-N
Synonym methyl phenoxyacetate, phenoxyacetic acid methyl ester, acetic acid, phenoxy-, methyl ester, methylphenoxyacetate, unii-jd9of3328o, phenoxy methyl acetate, methyl phenoxy acetate, phenoxyacetic acid methyl, acetic acid, 2-phenoxy-, methyl ester, pubchem12489
PubChem CID 16365
IUPAC Name methyl 2-phenoxyacetate
SMILES COC(=O)COC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 245°C
Quantity 500 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Phenoxyacetate