missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Pentadecanoate 98.0+%, TCI America™
$162.17 - $443.65
Chemical Identifiers
| CAS | 7132-64-1 |
|---|---|
| Molecular Formula | C16H32O2 |
| Molecular Weight (g/mol) | 256.43 |
| MDL Number | MFCD00008989 |
| InChI Key | XIUXKAZJZFLLDQ-UHFFFAOYSA-N |
| Synonym | pentadecanoic acid, methyl ester, pentadecanoic acid methyl ester, methyl n-pentadecanoate, unii-8k0zv6faiz, fatty acids, c12-18, me esters, 8k0zv6faiz, n-pentadecanoic acid methyl ester, methylpentadecanoate, pentadecanoic acid methyl, acmc-209oi7 |
| PubChem CID | 23518 |
| IUPAC Name | methyl pentadecanoate |
| SMILES | CCCCCCCCCCCCCCC(=O)OC |
Chemical Identifiers
| 7132-64-1 | |
| 256.43 | |
| XIUXKAZJZFLLDQ-UHFFFAOYSA-N | |
| 23518 | |
| CCCCCCCCCCCCCCC(=O)OC |
| C16H32O2 | |
| MFCD00008989 | |
| pentadecanoic acid, methyl ester, pentadecanoic acid methyl ester, methyl n-pentadecanoate, unii-8k0zv6faiz, fatty acids, c12-18, me esters, 8k0zv6faiz, n-pentadecanoic acid methyl ester, methylpentadecanoate, pentadecanoic acid methyl, acmc-209oi7 | |
| methyl pentadecanoate |
Specifications
| 7132-64-1 | |
| 155°C | |
| MFCD00008989 | |
| pentadecanoic acid, methyl ester, pentadecanoic acid methyl ester, methyl n-pentadecanoate, unii-8k0zv6faiz, fatty acids, c12-18, me esters, 8k0zv6faiz, n-pentadecanoic acid methyl ester, methylpentadecanoate, pentadecanoic acid methyl, acmc-209oi7 | |
| CCCCCCCCCCCCCCC(=O)OC | |
| 256.43 | |
| 256.43 | |
| Liquid |
| Yellow | |
| C16H32O2 | |
| 5 mL | |
| XIUXKAZJZFLLDQ-UHFFFAOYSA-N | |
| methyl pentadecanoate | |
| 23518 | |
| ≥98.0% (GC) | |
| Methyl Pentadecanoate |
Safety and Handling
TSCA : Yes