missing translation for 'onlineSavingsMsg'
Learn More

Methyl Palmitate 97.0+%, TCI America™

Catalog No. p-7134370
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000625G 25 g
P0006250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P000625G Supplier TCI America Supplier No. P000625G
Only null left

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Color White
Boiling Point 185°C
Quantity 25 g
Formula Weight 270.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Palmitate
EINECSNumber (2)-0798
TSCA Yes