missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 97%

CAS: 112-39-0 | C17H34O2 | 270.457 g/mol

Supplier:  Thermo Scientific Chemicals L0550936

 View more versions of this product

Catalog No. AAL0550936


Only null left

Description

Description

Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

Solubility
Soluble in ethanol, chloroform, dimethyl sulfoxide and dimethyl formamide. Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Methyl palmitate
28°C to 34°C
135°C to 137°C (0.2 mmHg)
C17H34O2
CH3(CH2)14CO2CH3
500 g
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
FLIACVVOZYBSBS-UHFFFAOYSA-N
methyl hexadecanoate
8181
270.46
112-39-0
0.852
>110°C (230°F)
1.4512
MFCD00008994
1780973
Soluble in ethanol,chloroform,dimethyl sulfoxide and dimethyl formamide. Insoluble in water.
CCCCCCCCCCCCCCCC(=O)OC
270.457
CHEBI:69187
97%

Chemical Identifiers

112-39-0
270.457
FLIACVVOZYBSBS-UHFFFAOYSA-N
8181
methyl hexadecanoate
C17H34O2
MFCD00008994
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
CHEBI:69187
CCCCCCCCCCCCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-966-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only