missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 97%

Catalog No. AAL0550922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0550922 100 g
AAL0550936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0550922 Supplier Thermo Scientific Chemicals Supplier No. L0550922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-39-0 | C17H34O2 | 270.457 g/mol

Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl palmitate is used in the preparation of detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and animal feeds. It exhibits an anti-inflammatory and anti-fibrotic agent and prevents bleomycin-induced lung inflammation and fibrosis in rats. In addition, it also prevents carbon tetrachloride-induced liver fibrosis linked to reduce transforming growth factor beta, which is a secreted protein that controls proliferation, cellular differentiation and other functions in most cells.

Solubility
Soluble in ethanol, chloroform, dimethyl sulfoxide and dimethyl formamide. Insoluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 28°C to 34°C
Density 0.852
Boiling Point 135°C to 137°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4512
Linear Formula CH3(CH2)14CO2CH3
Quantity 100 g
Beilstein 1780973
Solubility Information Soluble in ethanol,chloroform,dimethyl sulfoxide and dimethyl formamide. Insoluble in water.
Formula Weight 270.46
Percent Purity 97%
Chemical Name or Material Methyl palmitate
Show More Show Less
EINECSNumber 203-966-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only