missing translation for 'onlineSavingsMsg'
Learn More

Methyl palmitate, 95%

Catalog No. AC414685000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414685000 500 g Glass bottle
AC414681000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414685000 Supplier Thermo Scientific Chemicals Supplier No. 414685000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112-39-0 | C17H34O2 | 270.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
MDL Number MFCD00008994
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
PubChem CID 8181
ChEBI CHEBI:69187
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 28.0°C
Color Colorless
Density 0.85g/mL
Boiling Point 338.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)14CO2CH3
Quantity 500 g
Solubility Information Solubility in water: insoluble. Other solubilities: solubel in alcohol,ether
Formula Weight 270.45
Percent Purity 95%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl palmitate, 95%
Show More Show Less
EINECSNumber 203-966-3
TSCA TSCA

RUO – Research Use Only