missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfoxide, 98%

Catalog No. p-4089552
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428150010 1 g Glass bottle
AC428150050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428150010 Supplier Thermo Scientific Chemicals Supplier No. 428150010
Only null left

CAS: 934-72-5 | C8H10OS | 154.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 934-72-5
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00075175
InChI Key FEVALTJSQBFLEU-UHFFFAOYNA-N
Synonym 1-methyl-4-methylsulfinyl benzene, methyl 4-methylphenylsulfoxide, methyl p-tolyl sulfoxide, 1-methanesulfinyl-4-methylbenzene, methyl p-methylphenylsulfoxide, 1-methyl-4-methylsulphinyl benzene, benzene, 1-methyl-4-methylsulfinyl, acmc-20aotg, acmc-20apjq, methyl p-tolylsulfoxide
PubChem CID 136743
IUPAC Name 1-methyl-4-methylsulfinylbenzene
SMILES CC1=CC=C(C=C1)S(C)=O

Specifications

Melting Point 44°C to 46°C
Boiling Point 113°C to 114°C (2.2 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material Methyl p-tolyl sulfoxide

RUO – Research Use Only