missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfide, 97%

Catalog No. AAL0293306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0293306 5 g
AAL0293314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0293306 Supplier Thermo Scientific Chemicals Supplier No. L0293306

CAS: 623-13-2 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-13-2
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00008560
InChI Key VHILIAIEEYLJNA-UHFFFAOYSA-N
Synonym methyl p-tolyl sulfide, 4-methylthioanisole, 4-methylthio toluene, p-methylthio toluene, benzene, 1-methyl-4-methylthio, p-tolyl methyl sulfide, p-methylphenyl methyl sulfide, methyl p-cresyl sulfide, p-cresyl methyl sulfide
PubChem CID 69334
IUPAC Name 1-methyl-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C)C=C1

Specifications

Density 1.029
Boiling Point 81°C to 82°C (18 mmHg)
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.573
Quantity 5 g
UN Number UN3334
Beilstein 1905733
Formula Weight 138.23
Percent Purity 97%
Chemical Name or Material Methyl p-tolyl sulfide
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
EINECSNumber 210-773-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only